Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit

Catalog No. DCIM00
Change view
Click to view available options
Quantity:
1 Kit
6 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DCIM00 1 Kit
SCIM00 6 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DCIM00 Supplier R&D Systems™ Supplier No. DCIM00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ICAM-1/CD54 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).

Specifications

Assay Range 0.63 - 40ng/mL
Assay Sensitivity 0.053ng/mL
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules. < 50% cross-species reactivity observed with species tested.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human ICAM-1
Target Species Human
For Use With (Application) ELISA
Incubation Time 2.5 hr.
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.