Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ICAM-2 Fc Recombinant Protein, PeproTech®

Catalog No. 15022500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15022500UG 500 μg
15022100UG 100 μg
150-22-10UG 10 μg
150-22-1MG 1 mg
15022250UG 250 μg
150-22-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 15022500UG Supplier Gibco™ Supplier No. 15022500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO-derived Recombinant Human ICAM-2 Fc is a glycosylated, disulfide-linked homodimer of 435 amino-acid-residues whose monomer consists of the 202-amino-acid extracellular portion of ICAM-2 fused to the 231-amino-acid length Fc portion of human IgG by two glycine residues. The calculated molecular weight of monomeric CHO cell-derived Recombinant Human ICAM-2 Fc is 48.5 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of approximately 70-80 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13598
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3384
Molecular Weight (g/mol) 48.5 kDa
Name Human ICAM-2 Fc
Quantity 500 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001
Common Name ICAM-2
Gene Symbol ICAM2
Biological Activity Determined by its ability to support the adhesion of PMA-stimulated HSB2 cells to plates pre-coated with ICAM-3 Fc. The ED50 for this effect is 1.0-5.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence SDEKVFEVHV RPKKLAVEPK GSLEVNCSTT CNQPEVGGLE TSLDKILLDE QAQWKHYLVS NISHDTVLQC HFTCSGKQES MNSNVSVYQP PRQVILTLQP TLVAVGKSFT IECRVPTVEP LDSLTLFLFR GNETLHYETF GKAAPAPQEA TATFNSTADR EDGHRNFSCL AVLDLMSRGG NIFHKHSAPK MLEIYEPVSD SQGGPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.