Learn More
Sino Biological Human ICAM1 PAb 100UL 10346-T16-100
Supplier:  Sino Biological 10346T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ICAM-1/CD54 (rh ICAM-1/CD54; Catalog#10346-H08H; NP_000192.2; Met1-Glu480). ICAM-1/CD54 specific IgG was purified by Human ICAM-1/CD54 affinity chromatography.
Catalog No. 50-241-0823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.