Learn More
Sino Biological ICAM 5 / Intercellular adhesion molecule 5 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 16050T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ICAM 5 / Intercellular adhesion molecule 5 (rh ICAM 5 / Intercellular adhesion molecule 5; Catalog#16050-H08H; NP_003250.3; Met1-Pro834). ICAM 5 / Intercellular adhesion molecule 5 specific IgG was purified by Human ICAM 5 / Intercellular adhesion molecule 5 affinity chromatography.
Catalog No. 50-255-4067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.