Learn More
Sino Biological Human IDO1 / IDO Protein 50ug
Supplier:  Sino Biological 11650HNCE50
Encompass_Preferred
A DNA sequence encoding the human IDO1 (NP_002155.1) (Ala2-Gly403) was expressed and purified with one additional amino acids (Gly) at the N-terminus.
Catalog No. 50-224-7243
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.