Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IER5 Partial ORF (NP_057629, 1 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89948257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-257 10 μg
89-948-258 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-948-257 Supplier Abnova™ Supplier No. H00051278Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein that is similar to other immediate early response proteins. In the mouse, a similar gene may play an important role in mediating the cellular response to mitogenic signals. Studies in rats found the expression of a similar gene to be increased after waking and sleep deprivation. [provided by RefSeq]

Sequence: MEFKLEAHRIVSISLGKIYNSRVQRGGIKLHKNLLVSLVLRSARQVYLSDPCPGLY

Specifications

Accession Number NP_057629
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51278
Molecular Weight (g/mol) 31.9kDa
Name IER5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEFKLEAHRIVSISLGKIYNSRVQRGGIKLHKNLLVSLVLRSARQVYLSDPCPGLY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC102760/SBBI48
Common Name IER5
Gene Symbol IER5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.