Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IFIH1 Full-length ORF (AAH46208.1, 1 a.a. - 221 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89948281 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-281 10 μg
89-948-282 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-948-281 Supplier Abnova™ Supplier No. H00064135P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein that is upregulated in response to treatment with beta-interferon (IFNB) and a protein kinase C-activating compound, mezerein (MEZ). Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation. [provided by RefSeq]

Sequence: MSNGYSTDENFRYLISCFRARVKMYIQVEPVLDYLTFLPAEVKEQIQRTVATSGNMQAVELLLSTLEKGVWHLGWTREFVEALRRTGSPLAARYMNPELTDLPSPSFENAHDEYLQLLNLLQPTLVDKLLVRDVLDKCMEEELLTIEDRNRIAAAENNGNESGVRELLKRIVQKENWFSAFLNVLRQTGNNELVQELTGSDCSESNAGICNFTEEDSSNSA

Specifications

Accession Number AAH46208.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 64135
Molecular Weight (g/mol) 51.5kDa
Name IFIH1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MSNGYSTDENFRYLISCFRARVKMYIQVEPVLDYLTFLPAEVKEQIQRTVATSGNMQAVELLLSTLEKGVWHLGWTREFVEALRRTGSPLAARYMNPELTDLPSPSFENAHDEYLQLLNLLQPTLVDKLLVRDVLDKCMEEELLTIEDRNRIAAAENNGNESGVRELLKRIVQKENWFSAFLNVLRQTGNNELVQELTGSDCSESNAGICNFTEEDSSNSA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Hlcd/IDDM19/MDA-5/MDA5/MGC133047
Common Name IFIH1
Gene Symbol IFIH1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.