Learn More
Description
The Quantikine™ HS Human IFN-alpha 2 Immunoassay is a 4.0 hour solid phase ELISA designed to measure human IFN-alpha 2 (IFN-alpha 2a, IFN-alpha 2b, IFN-alpha 2c) in cell culture supernates, serum, and plasma. It contains HEK293-expressed recombinant human IFN-alpha 2a and has been shown to accurately quantitate the recombinant factor.
Results obtained using natural human IFN-alpha 2 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IFN-alpha 2.
Specifications
Specifications
| Assay Range | 0.78 to 50 pg/mL |
| Assay Sensitivity | 0.107 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and recombinant human IFN-alpha 2. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3440 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | alpha-2 A interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2 b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
