Learn More
ACROBiosystems ACROBiosystems Human IFN-alpha 2b (K46R) Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems IFBH5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IFN-alpha 2b (K46R), Fc Tag (IFB-H5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 188 (Accession # P01563-1 (K46R)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.7 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IFNAR2, His Tag (Cat. No. IF2-H5224) at 5 μg/mL (100 μL/well) can bind Human IFN-alpha 2b (K46R), Fc Tag (Cat. No. IFB-H5253) with a linear range of 2.4-10 ng/mL (QC tested).
Catalog No. 50-210-7260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.