Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IFN-alpha ELISA Kit
GSA_VASDP

Catalog No. 411002 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Plate
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
411002 5 Plates
411001 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 411002 Supplier R&D Systems™ Supplier No. 411002
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • Pre-coated microtiter plate(s)
  • Plate sealers
  • Wash Solution Concentrate
  • Human Interferon Alpha Standard, 10,000 pg/ml
  • Dilution Buffer
  • Antibody Concentrate
  • HRP Conjugate Concentrate

Human IFN-alpha ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IFN-alpha ELISA Kit quantifies human IFN-alpha in cell culture supernates (100 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 12.50 to 500 pg/mL (Cell Culture Supernates), 156.00 to 5000 pg/mL (Cell Culture Supernates)
Assay Sensitivity 12.5pg/mL
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant
Specificity Human IFN-alpha
Sufficient For 5 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3440
Quantity 5 Plates
Sample Volume Cell Culture Supernates (100 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.