Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8141SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8141SP 25 μg
MAB8141100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8141SP Supplier R&D Systems Supplier No. MAB8141SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization, Immunocytochemistry.

Specifications

Antigen IFN-beta
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 937912
Conjugate Unconjugated
Dilution Neutralization 0.2-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3456
Test Specificity Detects human IFN-beta in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.