Learn More
ACROBiosystems Human IFN-gamma 96tests
Supplier:  ACROBiosystems CFAC00196TESTS
Encompass_Preferred
The biology and function of IFN-γ has been extensively reviewed in the literature. Cytometric Bead Array (CBA) assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence making it possible to detect analytes using flow cytometry.
Catalog No. 50-253-5292
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.