Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IFN-gamma Quantikine ELISA Kit
SDP

Catalog No. DIF50C Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DIF50C 1 Kit
SIF50C 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DIF50C Supplier R&D Systems™ Supplier No. DIF50C

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

solid phase ELISA designed to measure human IFN-gamma levels in cell culture supernates, serum, and plasma.

Specifications

Assay Range 15.60 to 1000 pg/mL
Assay Sensitivity 5.69 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum
Specificity Natural and Recombinant Human IFN-gamma
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3458
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Target IFN-gamma
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.