Learn More
PBL Assay Science VeriKine-HS Human Interferon Alpha (IFN-Alpha, IFNa) All-Subtype ELISA, EDTA Plasma/Serum/Tissue Culture Media (TCM) (High Sensitivity) [1x96-well plate]
Supplier:  PBL Assay Science 411151
Encompass_Preferred
VeriKine-HS Human Interferon Alpha All Subtype ELISA Kit is manufactured by PBL Assay Science (Catalog #41115-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNa All Subtype ELISA is Serum, Plasma, and Tissue Culture Media (TCM) compatible, quantifies 12 of 12 human IFN-Alpha subtypes, and has an assay range of 1.95-125 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
Catalog No. 50-153-8060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.