Learn More
Description
Specifically detects IgE, Myeloma Clone: in samples, and it is validated for Isotype Control
Specifications
Specifications
| Antigen | IgE, Myeloma |
| Conjugate | Unconjugated |
| Formulation | In 150mM NaCl, 15mM potassium phosphate buffer, pH 7.4. |
| Host Species | Human |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Isotype Control (Primary) |
| Test Specificity | Native IgE from human myeloma plasma. |
| Content And Storage | −20°C |
| Form | Liquid |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.