Learn More
Abcam Human IgE Protein, Full Length, Unconjugated, 1 mg
Description
- Purity: >90%
- Endotoxin level: < 25 EU/mg
- Predicted molecular weight: 185 kDa
- Applications: FuncS, IHC, ELISA, SDS-PAGE, WB
- Form: Liquid
- Preservative: 0.09% Sodium azide
- Constituents: PBS, 0.88% Sodium chloride
- pH: 7.4
- Species: Human
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | IgE (Human) |
| Shipping Condition | Blue Ice |
| Substrate Properties | Protein-based Substrate |
| Reactivity | Human |
| Label or Dye | Unconjugated |
| Buffer | PBS, 0.88% sodium chloride, 0.09% sodium azide, pH7.4 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Cross Reactivity | No cross-reaction from other immunoglobulin isotypes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.