Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IGF-BP2 Recombinant Protein, PeproTech®

Catalog No. 35006B250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35006B250UG 250 μg
35006B100UG 100 μg
35-006-B01M 2 x 500 μg
35006B20UG 20 μg
35006B500UG 500 μg
35006B50UG 50 μg
350-06B-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 35006B250UG Supplier Gibco™ Supplier No. 35006B250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

350-06B-1MG will be provided as 2 x 500 μg (350-06B-500UG). Recombinant Human IGF-BP2 is a 31.5 kDa protein consisting of 289 amino acid residues including the IGF-BP domain and thyroglobulin type-I domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18065
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3485
Molecular Weight (g/mol) 31.5 kDa
Name Human IGF-BP2
Quantity 250 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36 kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753
Common Name IGFBP2
Gene Symbol IGFBP2
Biological Activity Determined by its ability to inhibit IGF-I induced proliferation of FDC-P1 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence EVLFRCPPCT PERLAACGPP PVAPPAAVAA VAGGARMPCA ELVREPGCGC CSVCARLEGE ACQVYTPRCG QGLRCYPHPG SELPLQALVM GEGTCEKRRD AEYGASPEQV ADNGDDHSEG GLVENHVDST MNMLGGGGSA GRKPLKSGMK ELAVFREKVT EQHRQMGKGG KHHLGLEEPK KLRPPPARTP CQQELDQVLE RISTMRLPDE RGPLEHLYSL HIPNCDKHGL YNLKQCKMSL NGQRGECWCV NPNTGKLIQG APTIRGDPEC HLFYNEQQEA RGVHTQRMQ
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.