Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IGF-BP6 Recombinant Protein, PeproTech®

Catalog No. 35007B5UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 250 μg
20 μg
250 μg
4 x 250 μg
5 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
350-07B-5UG 5 μg
35007B100UG 100 μg
35-007-B500 2 x 250 μg
35007B20UG 20 μg
35-007-B01M 4 x 250 μg
35007B250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 350-07B-5UG Supplier Gibco™ Supplier No. 35007B5UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

350-07B-1MG will be provided as 4 x 250 μg (350-07B-250UG). 350-07B-500UG will be provided as 2 x 250 μg (350-07B-250UG). RecombinantRecombinant Human IGF-BP6 migrates at an apparent molecular weight of approximately 23.0-30.0 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6, which specifically inhibits IGF-II actions, is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P24592
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3489
Molecular Weight (g/mol) 22.3 kDa
Name Human IGF-BP6
Quantity 5 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias IBP6; IBP-6; IGF binding protein 6; IGF-binding protein 6; IGFBP; IGFBP6; IGF-BP6; Igfbp-6; Insulin like growth factor binding protein; insulin like growth factor binding protein 6; insulin-like growth factor binding protein 6; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 6 PRECURSOR (IGFBP-6) (IBP-6) (IGF-BINDING PROTEIN 6); insulin-like growth factor-binding protein 6; RBP6A
Common Name IGFBP6
Gene Symbol IGFBP6
Biological Activity Determined by its ability to inhibit IGF-II induced proliferation of human MCF-7 cells. The expected ED50 for this effect is 0.1 -0.4 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence RCPGCGQGVQ AGCPGGCVEE EDGGSPAEGC AEAEGCLRRE GQECGVYTPN CAPGLQCHPP KDDEAPLRAL LLGRGRCLPA RAPAVAEENP KESKPQAGTA RPQDVNRRDQ QRNPGTSTTP SQPNSAGVQD TEMGPCRRHL DSVLQQLQTE VYRGAQTLYV PNCDHRGFYR KRQCRSSQGQ RRGPCWCVDR MGKSLPGSPD GNGSSSCPTG SSG
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.