Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IGF-BP7 Recombinant Protein, PeproTech®

Catalog No. 3500950UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
350-09-50UG 50 μg
35009100UG 100 μg
350-09-1MG 1 mg
35009250UG 250 μg
350-09-25UG 25 μg
35009500UG 500 μg
350-09-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 350-09-50UG Supplier Gibco™ Supplier No. 3500950UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IGF-BP7 is a 26.4 kDa protein consisting of 256 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of the insulin-like growth factor (IGF)-binding protein (IGFBP) family. IGFBPs bind IGFs with high affinity, and regulate IGF availability in body fluids and tissues and modulate IGF binding to its receptors. This protein binds IGF-I and IGF-II with relatively low affinity, and belongs to a subfamily of low-affinity IGFBPs. It also stimulates prostacyclin production and cell adhesion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene, and one variant has been associated with retinal arterial macroaneurysm.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16270
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3490
Molecular Weight (g/mol) 26.4 kDa
Name Human IGF-BP7
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AGM; angiomodulin; FSTL2; IBP7; IBP-7; IGF-binding protein 7; IGFBP; IGFBP rP1; Igfbp7; IGFBP-7; IGFBP-7 V; IGFBPRP1; IGFBP-rP1; Insulin like growth factor binding protein; insulin like growth factor binding protein 7; insulin-like growth factor binding protein 7; insulin-like growth factor-binding protein 7; Mac25; MAC25 protein; PGI2-stimulating factor; prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factor
Common Name IGFBP7
Gene Symbol IGFBP7
Conjugate Unconjugated
Recombinant Recombinant
Sequence SSSDTCGPCE PASCPPLPPL GCLLGETRDA CGCCPMCARG EGEPCGGGGA GRGYCAPGME CVKSRKRRKG KAGAAAGGPG VSGVCVCKSR YPVCGSDGTT YPSGCQLRAA SQRAESRGEK AITQVSKGTC EQGPSIVTPP KDIWNVTGAQ VYLSCEVIGI PTPVLIWNKV KRGHYGVQRT ELLPGDRDNL AIQTRGGPEK HEVTGWVLVS PLSKEDAGEY ECHASNSQGQ ASASAKITVV DALHEIPVKK GEGAEL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.