Learn More
R&D Systems™ Human IGF-I/IGF-1 Quantikine ELISA Kit

Description
- A 4.5 hour solid-phase ELISA designed to measure human IGF-1 in cell culture supernates, serum, and plasma.
- It contains E. coli-expressed recombinant human IGF-1 and has antibodies raised againt the recombinant protein.
- Results obtained for naturally occurring human IGF-1 showed linear curves that were parallel to the standard curves obtained using the Quantikine™ kit standards.
- These results indicate that this kit can be used to determine relative mass values for natural human IGF-1.
- Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested twenty times on one plate to assess intra-assay precision
- Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in twenty separate assays to assess inter-assay precision. Assays were performed by at least three technicians using two lots of components
Specifications
Specifications
| Assay Range | 0.06 - 4 ng/mL |
| Assay Sensitivity | 0.022 ng/mL |
| Product Type | ELISA |
| Sample Type | Plasma (EDTA), Heparin Plasma, Serum, Serum-free Cell Culture Supernatant |
| Specificity | Natural and Recombinant Human IGF-1 |
| Gene ID (Entrez) | 3479 |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.