Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IGF1R (NM_000875, 954- 1367 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells

Catalog No. 89948453 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-453 Supplier Abnova™ Supplier No. P4702
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

This receptor binds insulin-like growth factor with a high affinity. It has tyrosine kinase activity. The insulin-like growth factor I receptor plays a critical role in transformation events. Cleavage of the precursor generates alpha and beta subunits. It is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival. [provided by RefSeq]

Specifications

Accession Number NM_000875
Concentration 0.183 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 3480
Molecular Weight (g/mol) 76.532kDa
Name IGF1R (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method One-step affinity purification using GSH-agarose
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias CD221/IGFIR/JTK13/MGC142170/MGC142172/MGC18216
Common Name IGF1R
Gene Symbol IGF1R
Biological Activity 250pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST-His
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.