Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IGF2R ELISA Kit

Catalog No. EH251RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH251RB 96 Tests
EH251RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH251RB Supplier Invitrogen™ Supplier No. EH251RB
Add to Cart
Add to Cart

ELISA

Human IGF2R quantitates human IGF2R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IGF2R.

Lysosomal enzymes containing one or two mannose 6-phosphate (man6P) moieties are moved about in the cell by two distinct but interconnected cycles by means of approximately 300 kDa cation independent mannose 6-phospate receptors (CI-MPR). The biosynthetic cycle refers to the MPR transport of newly synthesized lysosomal enzymes from the trans Golgi network to late endosomes or early lysosomes. The endocytic cycle transports extracellular lysosomal enzymes from the plasma membrane via clathrin-coated vesicles to early endosomes. The entire pool of MPRs cycles between these cellular compartments every 3 hours. The steady state distribution of MPR's is predominantly within late endosomes, fewer in the trans Golgi network and approximately 10 % at cell surface. In addition to its man6P binding activity, the MPR contains a separate binding site for the type II insulin-like growth factor and is capable of binding both man6P and IGF-II simultaneously. An approximately 240 kDa soluble, truncated form, representing the extracellular domain of the protein, has also been found circulating in serum and is capable of binding both ligands.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 300 kDa mannose 6-phosphate receptor; AI661837; cation-independent mannose 6-phosphate receptor; cation-independent mannose 6-phosphate receptor protein; cation-independent mannose-6-phosphate receptor; cation-independent MPR; CD222; CI Man-6-P receptor; CI-M6PR; CIMPR; CI-MPR; Igf2r; IGF-II receptor; IGF-II/CI-MPR; Igfr2; insulin like growth factor 2 receptor; insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; insulin-like growth factor receptor type 2; M6P; M6P/IGF2 receptor; M6P/IGF2R; M6PR; M6P-R; mannose-6-phosphate receptor, cation independent; mannose-6-phosphate/insulin-like growth factor II receptor; MPR 300; MPR1; MPR300; MPRI; similar to mannose-6-phosphate/insulin-like growth factor II receptor
Assay Range 0.27 to 200 ng/mL
Assay Sensitivity 0.27 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3482
Gene Symbol IGF2R
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IGF2R
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.