Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IGFBP-3 Quantikine ELISA Kit
SDP

Catalog No. SGB300 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SGB300 6 Plates 6 x 96-well Plates
DGB300 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SGB300 Supplier R&D Systems™ Supplier No. SGB300
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human IGFBP-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IGFBP-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IGFBP-3 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.78 - 50ng/mL
Assay Sensitivity 0.14ng/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human IGFBP-3
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3486
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.