Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IGFBP-4 Recombinant Protein, PeproTech®

Catalog No. 35005B5UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 250 μg
20 μg
250 μg
4 x 250 μg
5 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
350-05B-5UG 5 μg
35005B100UG 100 μg
35-005-B01M 4 x 250 μg
35005B20UG 20 μg
35-005-B500 2 x 250 μg
35005B250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 350-05B-5UG Supplier Gibco™ Supplier No. 35005B5UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

350-05B-1MG will be provided as 4 x 250 μg (350-05B-250UG). 350-05B-500UG will be provided as 2 x 250 μg (350-05B-250UG). Recombinant Human IGF-BP4 is a 25.7 kDa protein consisting of 237 amino acid residues including, the IGF-BP domain and thyroglobulin type-I domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22692
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3487
Molecular Weight (g/mol) 25.7 kDa
Name Human IGFBP-4
Quantity 5 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI875747; Band 3 A; BP-4; Deb2; differentially expressed in B16F10 2; HT29-IGFBP; IBP4; IBP-4; IGF-binding protein 4; IGFBP; Igfbp4; IGF-BP4; IGFBP-4; Insulin like growth factor binding protein; insulin like growth factor binding protein 4; insulin-like growth factor binding protein 4; insulin-like growth factor binding protein-4; insulin-like growth factor-binding protein 4; insulin-like growth factor-binding protein-4; LOW QUALITY PROTEIN: insulin-like growth factor-binding protein 4
Common Name IGFBP4
Gene Symbol IGFBP4
Biological Activity Determined by its ability to inhibit IGF-I induced proliferation of FDC-P1 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence DEAIHCPPCS EEKLARCRPP VGCEELVREP GCGCCATCAL GLGMPCGVYT PRCGSGLRCY PPRGVEKPLH TLMHGQGVCM ELAEIEAIQE SLQPSDKDEG DHPNNSFSPC SAHDRRCLQK HFAKIRDRST SGGKMKVNGA PREDARPVPQ GSCQSELHRA LERLAASQSR THEDLYIIPI PNCDRNGNFH PKQCHPALDG QRGKCWCVDR KTGVKLPGGL EPKGELDCHQ LADSFRE
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.