Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IgG ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB112445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112445 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112445 Supplier Novus Biologicals™ Supplier No. NBP300400
Add to Cart
Add to Cart

Human IgG ELISA Kit (Chemiluminescence) measures IgG in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Human IgG in samples. No significant cross-reactivity or interference between Human IgG and analogues was observed.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3500
Gene Symbol IGHG1
Quantity 1 Kit
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Immunoglobulin G, ImmunoglobulinG
Target IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.