Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG Fc Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB110V025
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.Specifications
| IgG | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3500 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 97924 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region | |
| 25 Tests | |
| Primary | |
| Detects chimeric proteins containing a human IgG Fc region in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction