Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG Fc Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB110V100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB110V100 100 Tests
FAB110V025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB110V100 Supplier R&D Systems Supplier No. FAB110V100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG
Applications Flow Cytometry
Classification Monoclonal
Clone 97924
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3500
Test Specificity Detects chimeric proteins containing a human IgG Fc region in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.