Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG (H+L) Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. G101CABS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G101CABS 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. G101CABS Supplier Novus Biologicals Supplier No. G101CABS
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IgG Polyclonal specifically detects IgG in Human samples. It is validated for Secondary.

Specifications

Antigen IgG
Applications Not Available
Classification Polyclonal
Conjugate Unconjugated
Dilution Hidden
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with No Preservative
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Immunogen Human IgG
Purification Method Antigen Affinity-purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity This antibody reacts with heavy chains on human IgG and with light chains on all human immunoglobulins. No reactivity is observed to non-immunoglobulin human, mouse, rabbit or bovine serum proteins by IEP.
Reconstitution Reconstitute with sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.