Learn More
Invitrogen™ Human IgG Isotype Control
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity/Antibody Specificity: The precipitin lines against goat anti-human whole serum are the same as those against goat anti-human IgG, Fc fragment specific when assayed by immunoelectrophoresis. No cross-reactivity was observed against goat anti-human IgM, Fc5-mu fragment specific or goat anti-human IgA, alpha chain specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Storage: After opening, dilute only enough antibody for a single day's use. The human source material used to make this antibody has been tested and found negative for Anti-HIV-1/HIV-2),HCV antibodies, HBsAg and ALT (SGPT). Country of Origin: USA.
Specifications
Specifications
| Antigen | Human IgG |
| Applications | Control, Inhibition Assays, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 11.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Gene Alias | Immunoglobulin G; ImmunoglobulinG |
| Host Species | Human |
| Purification Method | Purified |
| Quantity | 10 mg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.