Learn More
Invitrogen™ Human IgG1 Chimeric Recombinant Isotype Control (MOPC-21)
Description
Specificity: An antibody of unknown specificity that maybe used as a negative control.
Specifications
Specifications
| Antigen | Human IgG1 Chimeric |
| Applications | Control |
| Classification | Recombinant Monoclonal |
| Clone | MOPC-21 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | NOTCH2 |
| Gene Accession No. | O35516, Q04721 |
| Gene Alias | IGHG1; immunoglobulin heavy constant gamma 1 (G1m marker) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.