Learn More
Raybiotech Inc Human IgG1 ELISA Kit
Supplier:  Raybiotech Inc ELHIGG11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 50 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Detect human IgG1. No cross reactivity was found againt IgG2, IgG3, IgG4, IgM, IgA, IgD and IgE
Catalog No. 50-194-9387
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.