Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Human IgG1 Expression Control Vector

Catalog No. MIR6250 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6250 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6250 Supplier Mirus Bio™ Supplier No. MIR 6250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Sufficient for transfecting up to 100 mL of suspension CHO or HEK 293 cells in culture

  • Contains mixture of codon-optimized, CMV-driven plasmids that express heavy and light chains of human IgG1
  • Provided as 1 mg/mL transfection-grade plasmid mixture

Specifications

Promoter CMV
Product Type Expression Control Vector
Content And Storage Store Human IgG1 Expression Control at -20°C
Cloning Method Restriction Enzyme/MCS
Concentration 100 μg at 1 mg/mL
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.