Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgG3 Monoclonal Antibody (KT133)
GREENER_CHOICE

Catalog No. PIMA549521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549521 Supplier Invitrogen™ Supplier No. MA549521
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product was used in both Indirect and Sandwich ELISA. This product does not cross-react with IgG1, IgG2, IgG4, IgA, IgM and IgE. This product can be paired with MA5-49581.

Human IgG3, also known as human immunoglobulin G3, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the least abundant subclass of IgG in the blood and plays a minor role in defense against pathogens. IgG3 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG3 is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG3 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone KT133
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Alias IgG3; IGHG3; immunoglobulin heavy constant gamma 3 (G3m marker)
Host Species Mouse
Immunogen Recombinant Human IgG3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.