Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG4 Antibody, R&D Systems™

Catalog No. p-7146636
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB98951SP 25 μg
MAB98951100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB98951SP Supplier R&D Systems Supplier No. MAB98951SP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Human IgG4 Monoclonal specifically detects Human IgG4 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Human IgG4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2350A
Conjugate Unconjugated
Dilution Western Blot 2 μg;/mL, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias Ig gamma-4 chain C region, immunoglobulin heavy constant gamma 4 (G4m marker), MGC117419
Host Species Rabbit
Immunogen Synthetic peptide containing Human IgG4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3503.0
Test Specificity Detects human IgG4 in Western blots and ELISA. In Western blots, no cross-reactivity with human IgG1, human IgG2, and human IgG3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.