Learn More
Sartorius Human IgGTiter & Viability Kit
Catalog No. 502891663 Shop All Sartorius Products
Encompass
iQue Human IgG Titer & Viability is the only multiplexed assay to enable simultaneous reporting of IgG quantity cell number and cell viability in each well of cell line production screening plates. These metrics enable the precise calculation of the IgG quantity per cell and per viable cell to profile cell productivity. The Only Solution Enjoy expedited workflows with the iQue Human IgG Titer & Viability for use in flow cytometry applications. The iQue Human IgG Titer & Viability is the only solution that correlates Immunoglobulin G IgG quantitation per cell health and growth readouts in a single cost-effective competition assay using IgG-capture beads IgG fluorescent competing antibody and a cell viability stain. Take advantage of this popular unique kit.
Catalog No. 50-289-1663 Supplier Sartorius Supplier No. 91142
iQue Human IgG Titer & Viability is the only multiplexed assay to enable simultaneous reporting of IgG quantity cell number and cell viability in each well of cell line production screening plates. These metrics enable the precise calculation of the IgG quantity per cell and per viable cell to profile cell productivity. The Only Solution Enjoy expedited workflows with the iQue Human IgG Titer & Viability for use in flow cytometry applications. The iQue Human IgG Titer & Viability is the only solution that correlates Immunoglobulin G IgG quantitation per cell health and growth readouts in a single cost-effective competition assay using IgG-capture beads IgG fluorescent competing antibody and a cell viability stain. Take advantage of this popular unique kit.