Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human IgM, Myeloma, Isotype Control,

Catalog No. 4011081MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-110-81MG 1 mg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-110-81MG Supplier MilliporeSigma™ Supplier No. 4011081MG
Add to Cart
Add to Cart

Human Isotype Control Antibody

Specifically detects IgM, Myeloma Clone: in samples, and it is validated for Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM, Myeloma
Conjugate Unconjugated
Formulation In 200mM NaCl, 50mM Tris, pH 8.0.
Host Species Human
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Test Specificity Native IgM from human myeloma plasma.
Content And Storage −20°C
Form Liquid
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.