Learn More
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Immunohistochemistry, SDS-PAGE |
| Formulation | 0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0 |
| Quantity | 1 mg |
| Storage Requirements | Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze/thaw cycles. |
| Gene Alias | AGM1, MU, VH |
| Inhibitor Properties | This product has been tested in SDS-Page and can be used as a control or standard in indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting. |
| Product Line | Novus |
| Purity or Quality Grade | Multi-step |
| Protein | Human IgM |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
