Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgM ProQuantum Immunoassay Kit

Catalog No. A45249
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A45249 96 Tests
A452495 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A45249 Supplier Invitrogen™ Supplier No. A45249
Add to Cart
Add to Cart

ELISA

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01871
Assay Range 0.0032 to 250 ng/mL
Assay Sensitivity <0.01 ng/mL
Conjugate FAM™
Product Type ProQuantum
Sample Type Plasma, Serum
For Use With (Equipment) qPCR Instruments
Gene ID (Entrez) 3507
Gene Symbol IGHM
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL
Storage Requirements -20°C
Target Human IgM
Test Time 30 mins
Total Assay Time 2 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.