Learn More
Sino Biological Human IKBKB PAb 100UL 101200-T08-100
Supplier:  Sino Biological 101200T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human IKBKB / IKK beta, and purified by antigen affinity chromatography.
Catalog No. 50-240-9909
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.