Learn More
ACROBiosystems Biotinylated Human IL-1 alpha, His,Avitag (ILA-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 113 - Ala 271 (Accession # P01583-1).
Supplier:  ACROBiosystems ILAH82E725UG
Encompass_Preferred
Interleukin-1 alpha (IL1a) is also known as IL-1A, IL1, IL1-ALPHA,IL1F1,and is a cytokine of the interleukin-1 family. IL1a possesses a wide spectrum of metabolic, physiological, haematopoietic activities, and plays one of the central roles in the regulation of the immune responses. It binds to the interleukin-1 receptor. IL-1α is constitutively produced by epithelial cells. It is found in substantial amounts in normal human epidermis and is distributed in a 1:1 ratio between living epidermal cells and stratum corneum.The constitutive production of large amounts of IL-1α precursor by healthy epidermal keratinocytes interfere with the important role of IL-1α in immune responses, assuming skin as a barrier, which prevents the entry of pathogenic microorganisms into the body. In vitro, IL-1α possesses biological effect on cells in the picomolar to femtomolar range. In vivo, Shortly after an onset of an infection into organism, IL-1α activates a set of immune system response processes.
Catalog No. 50-223-0689
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.