Learn More
Human IL-1 beta/IL-1F2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IL-1 beta/IL-1F2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1027B |
| Conjugate | Unconjugated |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P01584 |
| Gene Alias | catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.