Learn More
ACROBiosystems ACROBiosystems Human IL-1 beta / IL-1F2 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems ILBH51H350UG
Encompass_Preferred
Recombinant Protein;50UG;Human IL-1 beta, His Tag (ILB-H51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # P01584-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 19-20 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IL-1 beta, His Tag (Cat. No. ILB-H51H3) at 10 μg/mL (100 μL/well) can bind Human IL-1 RII, Fc Tag (Cat. No. IL2-H4256) with a linear range of 2-13 ng/mL (QC tested).
Catalog No. 50-210-7067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.