Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-1 R3/IL-1 R Acp ELISA Kit

Catalog No. EH255RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH255RB 96 Tests
EH255RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH255RB Supplier Invitrogen™ Supplier No. EH255RB
Add to Cart
Add to Cart

ELISA

Human IL-1 R3/IL-1 R Acp quantitates human IL-1 R3/IL-1 R Acp in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-1 R3/IL-1 R Acp.

This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1 R 3; IL 1R3; IL 1 RAcP; IL 1 RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1 R-3; IL1RAcP; IL-1 RAcP; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain
Assay Range 18 to 4,000 pg/mL
Assay Sensitivity 18 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3556
Gene Symbol IL1RAP
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL-1 RAcP
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.