Learn More
R&D Systems™ Human IL1RAPL1 DuoSet ELISA
Description
Specifications
Specifications
| Assay Range | 62.50 - 4000pg/mL |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 11141 |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | IL1R8IL1RAPL, IL-1R9, IL-1 RAPL1, IL-1-RAPL-1, IL-1 RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MR x 10, MR x 21, MR x 34, oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2, TIGIRR-2 mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 |
| Target | IL1RAPL1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.