Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the estimation of interleukin 10 (IL-10) in human serum and plasma, tissue homogenates and other biological fluids. Assay Range: 7.8 pg/ml - 10 ng/ml; Sensitivity: ~1.34 pg/ml.
Supplier:  KRISHGEN BIOSYSTEMS KB1072
Encompass_Preferred
Pack Size: 1 x 96 wells. This ELISA uses a sandwich assay format for better sensitivity. Introduction: Interleukin-10 (IL-10), also known as human cytokine synthesis inhibitory factor (CSIF), is an antiinflammatory cytokine. This cytokine is produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II antigens, and stimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747145
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.