Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-10 (ELISA Standard) Recombinant Protein

Catalog No. 501129335
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9335 4 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-9335 Supplier Invitrogen™ Supplier No. 29810965
Add to Cart
Add to Cart

Recombinant Protein

Description: Human IL-10 is a ~18 kDa factor also known as cytokine synthesis inhibitory factor (CSIF). Th2 T cells, B1 B cells and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells and acts as a costimulatory signal for mast cells, developing thymocytes and Th2 response. Applications Reported: Human IL-10 RSG ELISA Lyophilized Standard has been reported for use in the Human IL-10 ELISA RSG (Product # 88-7106). Applications Tested: The intended use of the Human IL-10 RSG ELISA Lyophilized Standard is as a standard for the Human IL-10 ELISA (Product # 88-7106-77). Typically the standard is run in a series of 7 doubling dilutions from the 'top' standard. Please refer to the Certificate of Analysis included with each ELISA set for instructions on the proper reconstitution and dilution of the lyophilized standard. Note: These are single-use vials. Use one time within 4 hours of reconstitution and discard. Reconstitute and dilute as suggested in the Certificate of Analysis or recovery may not be accurate. Source: E. coli expressed amino acids Ser 19-Asn 178 of mature human IL-10 accession # NM_000572.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22301
For Use With (Application) Neutralization, ELISA Standard
Formulation Protein with no preservative
Gene ID (Entrez) 3586
Molecular Weight (g/mol) 18 kDa
Name Human IL-10 (ELISA Standard)
Purification Method Purified
Quantity 4 pack
Source E. Coli
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Common Name IL-10
Gene Symbol IL10
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human IL-10, amino acids Ser19-Asn178 (Accession # NM_000572)
Content And Storage 4°C
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.