Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-10 R beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067972
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB874SP 25 μg
MAB874100 100 μg
MAB874500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB874SP Supplier R&D Systems Supplier No. MAB874SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

IL-10 R beta Monoclonal specifically detects IL-10 R beta in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.

Specifications

Antigen IL-10 R beta
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 90220
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.5-2 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q08334
Gene Alias CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4
Gene Symbols IL10RB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-10 R beta Met20-Ser220 Accession # Q08334
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3588
Test Specificity Detects human IL-10 R beta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10 R alpha , rhIFN-gamma RI, and rhIFN-gamma R2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.