Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-11 Quantikine ELISA Kit
SDP

Catalog No. D1100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1100 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D1100 Supplier R&D Systems™ Supplier No. D1100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human IL-11 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-11 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-11 in cell culture supernates (10 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).

Specifications

Conjugate HRP
Quantity 1 Plate
Sensitivity 8 pg/mL
Packaging 96-well Strip Plate
Synonym Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11 Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) ELISA
Product Type Solid Phase Sandwich ELISA
Specificity Natural and recombinant human IL-11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.