Learn More
Human IL-11 R alpha Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IL-11R alpha |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1020854 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q14626 |
| Gene Alias | AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.