Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-12 p70 Quantikine HS ELISA Kit

Catalog No. HS120
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HS120 1 Plate
SS120 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. HS120 Supplier R&D Systems™ Supplier No. HS120
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human IL-12 p70 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-12 p70 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-12 p70/IL-12A in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul).

Specifications

Assay Range 0.63 to 40 pg/mL (Cell Culture Supernates), 0.78 to 50 pg/mL (Serum, Heparin Plasma, EDTA Plasma)
Assay Sensitivity 0.5pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human IL-12p70
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3592
Quantity 1 Plate
Sample Volume Cell Culture Supernates (200uL),Serum (200uL),EDTA Plasma (200uL),Heparin Plasma (200uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12 A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.